accessibility menu, dialog, popup

UserName

CAS RN 871329-61-2

BSOONOHOH

CAS RN 871329-61-2

IUPAC Name: [3-(pyrrolidine-1-sulfonyl)phenyl]boronic acid
Synonyms: 3-(pyrrolidine-1-sulfonyl)phenylboronic acid
Molecular Weight (g/mol): 255.1
Molecular Formula: C10H14BNO4S
InChi Key: RCRUXZHMXFQVRF-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)S(=O)(=O)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results