accessibility menu, dialog, popup

UserName

CAS RN 870281-83-7

NHOFNOO

CAS RN 870281-83-7

IUPAC Name: 2-fluoro-6-nitro-N-phenylbenzamide
Synonyms: 2-fluoro-6-nitro-N-phenylbenzamide
Molecular Weight (g/mol): 260.22
Molecular Formula: C13H9FN2O3
InChi Key: GQVRPSFDUVNMDA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C(=O)NC2=CC=CC=C2)C(F)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results