accessibility menu, dialog, popup

UserName

H3COOOHOHOOCH3

CAS RN 87-91-2

IUPAC Name: 1,4-diethyl 2,3-dihydroxybutanedioate
Synonyms: diethyl tartrate
Molecular Weight (g/mol): 206.19
Molecular Formula: C8H14O6
InChi Key: YSAVZVORKRDODB-UHFFFAOYNA-N
SMILES: CCOC(=O)C(O)C(O)C(=O)OCC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (4)
  • (2)
  • (2)