accessibility menu, dialog, popup

UserName

CAS RN 87-72-9

IUPAC Name: (3S,4R,5R)-oxane-2,3,4,5-tetrol
Synonyms: D-arabinopyranose
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: SRBFZHDQGSBBOR-ZRMNMSDTSA-N
SMILES: O[C@@H]1COC(O)[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (3)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)