accessibility menu, dialog, popup

UserName

CAS RN 87-67-2

CH3NCH3CH3HOOH(R)(R)OHOOOHO

CAS RN 87-67-2

IUPAC Name: (2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
Synonyms: choline bitartrate
Molecular Weight (g/mol): 253.25
Molecular Formula: C9H19NO7
InChi Key: QWJSAWXRUVVRLH-LREBCSMRSA-M
SMILES: C[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (6)
  • (3)
  • (2)