accessibility menu, dialog, popup

UserName

CAS RN 869901-07-5

NNOH

CAS RN 869901-07-5

IUPAC Name: [6-(piperidin-1-yl)pyridin-2-yl]methanol
Synonyms: [6-(piperidin-1-yl)pyridin-2-yl]methanol
Molecular Weight (g/mol): 192.26
Molecular Formula: C11H16N2O
InChi Key: WLHPFWHASVOLSO-UHFFFAOYSA-N
SMILES: OCC1=NC(=CC=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results