accessibility menu, dialog, popup

UserName

CAS RN 86891-03-4

CH3OCH3ONa(E/Z)

CAS RN 86891-03-4

IUPAC Name: sodium 4-oxopent-2-en-2-olate hydrate
Synonyms: sodium 4-oxopent-2-en-2-olate hydrate
Molecular Weight (g/mol): 140.11
Molecular Formula: C5H9NaO3
InChi Key: UKDRZWCRRJOZNR-UHFFFAOYSA-M
SMILES: O.[Na+].CC([O-])=CC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results