CAS RN 86884-90-4
CAS RN 86884-90-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-2-Methyl Glycidol (>90%), TRC
CAS: 86884-90-4 Molecular Formula: C4H8O2 Molecular Weight (g/mol): 88.11 Synonym: Delamanid Impurity 9 IUPAC Name: [(2S)-2-methyloxiran-2-yl]methanol SMILES: C[C@]1(CO)CO1