accessibility menu, dialog, popup

UserName

CAS RN 868755-64-0

OSO

CAS RN 868755-64-0

IUPAC Name: 5-(furan-2-yl)thiophene-2-carbaldehyde
Synonyms: 5-(furan-2-yl)thiophene-2-carbaldehyde
Molecular Weight (g/mol): 178.21
Molecular Formula: C9H6O2S
InChi Key: IXUVFAGSDDBEKJ-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(S1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results