accessibility menu, dialog, popup

UserName

CAS RN 86845-28-5

CH3BrFFF

CAS RN 86845-28-5

IUPAC Name: 1-bromo-3-methyl-5-(trifluoromethyl)benzene
Synonyms: 1-bromo-3-methyl-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 239.04
Molecular Formula: C8H6BrF3
InChi Key: ZDAIYLXHTGIUGY-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(Br)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results