accessibility menu, dialog, popup

UserName

CAS RN 865471-20-1

ONFFFOOH

CAS RN 865471-20-1

IUPAC Name: 2-(morpholin-4-yl)-5-(trifluoromethyl)benzoic acid
Synonyms: 2-(morpholin-4-yl)-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 275.23
Molecular Formula: C12H12F3NO3
InChi Key: PRECFTDWCNEEDB-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=CC(=C1)C(F)(F)F)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results