CAS RN 865285-29-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
KKL-35, MedChemExpress
MedChemExpress KKL-35 is a trans-translation tagging reaction inhibitor with an IC50 of 0.9 μM.