accessibility menu, dialog, popup

UserName

OHFFFFFFFFFFFFFFFFFFFFF

CAS RN 865-86-1

IUPAC Name: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecan-1-ol
Synonyms: 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecan-1-ol
Molecular Weight (g/mol): 564.14
Molecular Formula: C12H5F21O
InChi Key: FLXYIZWPNQYPIT-UHFFFAOYSA-N
SMILES: OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)