accessibility menu, dialog, popup

UserName

KH3CCH3CH3O

CAS RN 865-47-4

IUPAC Name: potassium 2-methylpropan-2-olate
Synonyms: potassium tert-butoxide
Molecular Weight (g/mol): 112.21
Molecular Formula: C4H9KO
InChi Key: LPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES: [K+].CC(C)(C)[O-]
Molecular Weight (g/mol) 
  • (17)
  • (6)
Quantity 
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Grade 
  • (12)
  • (2)
Physical Form 
  • (5)
  • (7)
Percent Purity 
  • (5)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (16)
  • (5)
  • (17)
  • (6)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (12)
  • (2)
  • (5)
  • (7)
  • (5)
  • (5)