accessibility menu, dialog, popup

UserName

CAS RN 865-35-0

H3COCH3OCH3OCH3OCH3OTa

CAS RN 865-35-0

IUPAC Name: tantalum(5+) pentamethanolate
Synonyms: tantalum(5+) pentakis(methoxide)
Molecular Weight (g/mol): 336.12
Molecular Formula: C5H15O5Ta
InChi Key: QASMZJKUEABJNR-UHFFFAOYSA-N
SMILES: [Ta+5].C[O-].C[O-].C[O-].C[O-].C[O-]
Boiling Point 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)