CAS RN 86445-22-9
CAS RN 86445-22-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Promotions Available
Tocris Bioscience™ Auxinole
Tocris Bioscience™ Auxinole
Auxin antagonist of TIR1/AFB receptors; also OsTIR1 inhibitor in auxin-inducible degron (AID) system
2
Auxinole, TRC
CAS: 86445-22-9 Molecular Formula: C20H19NO3 Molecular Weight (g/mol): 321.37 Synonym: alpha-[2-(2,4-Dimethylphenyl)-2-oxoethyl]-1H-Indole-3-acetic Acid; IUPAC Name: 4-(2,4-dimethylphenyl)-2-(1H-indol-3-yl)-4-oxobutanoic acid SMILES: Cc1ccc(C(=O)CC(C(=O)O)c2c[nH]c3ccccc23)c(C)c1
3
Auxinole, MedChemExpress
MedChemExpress Auxinole is a potent auxin antagonist of TIR1/AFB receptors, binding TIR1 to block the formation of the TIR1-IAA-Aux/IAA complex and so inhibits auxin-responsive gene expression.