accessibility menu, dialog, popup

UserName

CAS RN 864068-79-1

CH3H3CCH3OONNSOH

CAS RN 864068-79-1

IUPAC Name: tert-butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate
Synonyms: tert-butyl 4-[4-(hydroxymethyl)-1,3-thiazol-2-yl]piperidine-1-carboxylate
Molecular Weight (g/mol): 298.4
Molecular Formula: C14H22N2O3S
InChi Key: RDZWXODJIWMJPE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)C1=NC(CO)=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results