accessibility menu, dialog, popup

UserName

CAS RN 863991-96-2

NOOHNOOHH2OHClHCl

CAS RN 863991-96-2

IUPAC Name: bis(3-[(piperidin-1-yl)methyl]benzoic acid) hydrate dihydrochloride
Synonyms: bis(3-(piperidin-1-ylmethyl)benzoic acid) hydrate dihydrochloride
Molecular Weight (g/mol): 529.5
Molecular Formula: C26H38Cl2N2O5
InChi Key: HTWIFQGKFFGIFX-UHFFFAOYSA-N
SMILES: O.Cl.Cl.OC(=O)C1=CC(CN2CCCCC2)=CC=C1.OC(=O)C1=CC(CN2CCCCC2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results