CAS RN 863598-09-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Casein Kinase II Inhibitor IV, MedChemExpress
MedChemExpress Casein Kinase II Inhibitor IV is a potent, ATP-competitive of casein kinase II inhibitor with an IC50 of 9 nM. Casein Kinase II Inhibitor IV ia also a human keratinocytes (NHEK) differentiation inducer.