CAS RN 86321-24-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ferulic Acid 4-O-beta-D-Glucuronide Disodium Salt, TRC
CAS: 86321-24-6 Molecular Formula: C16H18O10 Molecular Weight (g/mol): 370.31 Synonym: Ferulic Acid-B-O-B-D-Glucuronide SMILES: [Na+].[Na+].COc1cc(C=CC(=O)[O-])ccc1O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)[O-]