CAS RN 862543-54-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Maropitant Citrate Salt, TRC
CAS: 862543-54-2 Molecular Formula: C32 H40 N2 O . C6 H8 O7 Molecular Weight (g/mol): 660.8 Synonym: (2S-cis)-N-[[5-(1,1-Dimethylethyl)-2-methoxyphenyl]methyl]-2-(diphenylmethyl)-1-azabicyclo[2.2.2]octan-3-amine Citrate Salt,(2S,3S)-2-Benzhydryl-N-(5-tert-butyl-2-methoxybenzyl)quinuclidin-3-amine Citrate Salt,Cerenia SMILES: CC(C)(C)C1=CC(=C(C=C1)OC)CN[C@@H]2[C@@H](N3CCC2CC3)C(C4=CC=CC=C4)C5=CC=CC=C5.C(C(=O)O)C(CC(=O)O)(C(=O)O)O