accessibility menu, dialog, popup

UserName

CAS RN 862500-24-1

ONSOOO

CAS RN 862500-24-1

IUPAC Name: 2-(morpholine-4-sulfonyl)benzaldehyde
Synonyms: 2-(morpholine-4-sulfonyl)benzaldehyde
Molecular Weight (g/mol): 255.29
Molecular Formula: C11H13NO4S
InChi Key: GSDGTTZDQFKUBA-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1S(=O)(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results