accessibility menu, dialog, popup

UserName

CAS RN 86227-47-6

H3COOCH3(E)(E)(E)(E)(E)

CAS RN 86227-47-6

IUPAC Name: ethyl (5E,8E,11E,14E,17E)-icosa-5,8,11,14,17-pentaenoate
Synonyms: ethyl eicosapentaenoic acid
Molecular Weight (g/mol): 330.51
Molecular Formula: C22H34O2
InChi Key: SSQPWTVBQMWLSZ-LRKAYDMASA-N
SMILES: CCOC(=O)CCC\C=C\C\C=C\C\C=C\C\C=C\C\C=C\CC
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results