accessibility menu, dialog, popup

UserName

CAS RN 86176-56-9

IUPAC Name: 4-chloro-6-(2,5-dihydro-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
Synonyms: 4-chloro-6-(2H-1,2-oxazol-5-ylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 195.6
Molecular Formula: C9H6ClNO2
InChi Key: QCDHHUJPNPOGKY-UHFFFAOYSA-N
SMILES: ClC1=CC(=C2ONC=C2)C(=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results