accessibility menu, dialog, popup

UserName

CAS RN 86168-78-7

H3C(S)H3C(S)NHO(S)CH3HNO(S)OHONHO(S)CH3HNO(S)H2NHOOHN(S)ONH(S)(R)H3CHOOHN(S)H2NOONH(S)HOOHN(S)HOOHN(S)NHH2NNHONH(S)H2NOHN(S)CH3H3CONH(S)CH3H3COHNONH(S)H2NOOHN(S)CH3CH3ONH(S)OHOHN(S)CH3ONH(S)HNNH2HNOHN(S)NH2ONH(S)CH3H3COHN(S)CH3CH3ONH(S)H2NOOHN(S)OHOONH(S)(S)H3CH3COHN(S)SCH3ONH(S)OHOHN(S)NHNH2HNNH2OCH3OHO

CAS RN 86168-78-7

IUPAC Name: (3S)-3-{[(1S)-1-{[(1S,2S)-1-{[(1S)-1-{[(1S,2R)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-[({[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-5-amino-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-1-{[(1S,2S)-1-{[(1S)-1-{[(1S)-1-{[(1S)-4-carbamimidamido-1-carbamoylbutyl]carbamoyl}-2-hydroxyethyl]carbamoyl}-3-(methylsulfanyl)propyl]carbamoyl}-2-methylbutyl]carbamoyl}-2-carboxyethyl]carbamoyl}-3-carbamoylpropyl]carbamoyl}-3-methylbutyl]carbamoyl}-3-methylbutyl]carbamoyl}pentyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}ethyl]carbamoyl}-2-hydroxyethyl]carbamoyl}-3-methylbutyl]carbamoyl}-3-carbamoylpropyl]carbamoyl}methyl)carbamoyl]-3-methylbutyl]carbamoyl}-2-methylpropyl]carbamoyl}pentyl]carbamoyl}-4-carbamimidamidobutyl]carbamoyl}-2-(4-hydroxyphenyl)ethyl]carbamoyl}-2-hydroxyethyl]carbamoyl}-2-carbamoylethyl]carbamoyl}-2-hydroxypropyl]carbamoyl}-2-phenylethyl]carbamoyl}-2-methylbutyl]carbamoyl}ethyl]carbamoyl}-3-[(2S)-2-[(2S)-2-amino-3-(4-hydroxyphenyl)propanamido]propanamido]propanoic acid; acetic acid
Synonyms: sermorelin
Molecular Weight (g/mol): 3417.99
Molecular Formula: C151H250N44O44S
InChi Key: BVLCEKWPOSAKSZ-YQMCHIOTSA-N
SMILES: CC(O)=O.CC[C@H](C)[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](C)NC(=O)[C@@H](N)CC1=CC=C(O)C=C1)C(=O)N[C@@H](CC1=CC=CC=C1)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC1=CC=C(O)C=C1)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)NCC(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCNC(N)=N)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)