CAS RN 861139-16-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
RSV-IN-1, MedChemExpress
MedChemExpress RSV-IN-1 is a human respiratory syncytical virus (hRSV) inhibitor, with an IC50 of 0.11 μM.