accessibility menu, dialog, popup

UserName

CAS RN 86087-23-2

OOH

CAS RN 86087-23-2

IUPAC Name: oxolan-3-ol
Synonyms: 3-hydroxytetrahydrofuran
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: XDPCNPCKDGQBAN-UHFFFAOYNA-N
SMILES: OC1CCOC1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)