accessibility menu, dialog, popup

UserName

OONH(S)OHONHOO

CAS RN 86060-82-4

IUPAC Name: 6-{[(benzyloxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
Synonyms: 6-{[(benzyloxy)carbonyl]amino}-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 502.57
Molecular Formula: C29H30N2O6
InChi Key: KRULQRVJXQQPQH-UHFFFAOYNA-N
SMILES: OC(=O)C(CCCCNC(=O)OCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results