accessibility menu, dialog, popup

UserName

OCN

CAS RN 86-84-0

IUPAC Name: 1-isocyanatonaphthalene
Synonyms: naphthalene, 1-isocyanato-
Molecular Weight (g/mol): 169.18
Molecular Formula: C11H7NO
InChi Key: BDQNKCYCTYYMAA-UHFFFAOYSA-N
SMILES: O=C=NC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)