accessibility menu, dialog, popup

UserName

CAS RN 86-73-7

CAS RN 86-73-7

IUPAC Name: 9H-fluorene
Synonyms: fluorene o-biphenylenemethane diphenylenemethane methane, diphenylene- 2,3-benzindene diphenylene-methane 2,2'-methylenebiphenyl ccris 947
Molecular Weight (g/mol): 166.22
Molecular Formula: C13H10
InChi Key: NIHNNTQXNPWCJQ-UHFFFAOYSA-N
SMILES: C1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Concentration 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)