accessibility menu, dialog, popup

UserName

HOBOHN

CAS RN 86-58-8

IUPAC Name: (quinolin-8-yl)boronic acid
Synonyms: 8-quinolineboronic acid
Molecular Weight (g/mol): 172.98
Molecular Formula: C9H8BNO2
InChi Key: KXJJSKYICDAICD-UHFFFAOYSA-N
SMILES: OB(O)C1=C2N=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)