accessibility menu, dialog, popup

UserName

Cl

CAS RN 86-52-2

IUPAC Name: 1-(chloromethyl)naphthalene
Synonyms: naphthalene, 1-(chloromethyl)-
Molecular Weight (g/mol): 176.64
Molecular Formula: C11H9Cl
InChi Key: XMWGTKZEDLCVIG-UHFFFAOYSA-N
SMILES: ClCC1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (6)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (6)
  • (3)