accessibility menu, dialog, popup

UserName

CAS RN 859855-53-1

NH2NF

CAS RN 859855-53-1

IUPAC Name: 3-amino-4-fluorobenzonitrile
Synonyms: 3-amino-4-fluorobenzonitrile
Molecular Weight (g/mol): 136.13
Molecular Formula: C7H5FN2
InChi Key: BHMZMZOMQYFFQL-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results