accessibility menu, dialog, popup

UserName

CAS RN 859851-02-8

OSON

CAS RN 859851-02-8

IUPAC Name: 2H,3H-thieno[3,4-b][1,4]dioxine-5-carbonitrile
Synonyms: 2H,3H-thieno[3,4-b][1,4]dioxine-5-carbonitrile
Molecular Weight (g/mol): 167.18
Molecular Formula: C7H5NO2S
InChi Key: HSQAPDOVCIFQCU-UHFFFAOYSA-N
SMILES: N#CC1=C2OCCOC2=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results