accessibility menu, dialog, popup

UserName

CAS RN 859851-00-6

CH3NNOOOH

CAS RN 859851-00-6

IUPAC Name: 3-(furan-2-yl)-1-methyl-1H-pyrazole-5-carboxylic acid
Synonyms: 5-(furan-2-yl)-2-methylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 192.17
Molecular Formula: C9H8N2O3
InChi Key: MOPCEJWYTICWJM-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C(O)=O)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results