accessibility menu, dialog, popup

UserName

CAS RN 859850-99-0

ClNSO

CAS RN 859850-99-0

IUPAC Name: 2-(3-chlorophenyl)-1,3-thiazole-4-carbaldehyde
Synonyms: 2-(3-chlorophenyl)-1,3-thiazole-4-carbaldehyde
Molecular Weight (g/mol): 223.67
Molecular Formula: C10H6ClNOS
InChi Key: XMXWYXGIKFBCGR-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C1=NC(C=O)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results