CAS RN 859850-88-7
CAS RN 859850-88-7
IUPAC Name:
3-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
Synonyms:
3-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
Molecular Weight (g/mol):
218.3
Molecular Formula:
C13H18N2O
InChi Key:
KKYYCUZQPBFXKD-UHFFFAOYSA-N
SMILES:
CN1CCN(CC2=CC=CC(C=O)=C2)CC1
1
–
1
of
1
results
1
3-[(4-Methylpiperazin-1-yl)methyl]benzaldehyde, ≥95%, Thermo Scientific™
CAS: 859850-88-7 Molecular Formula: C13H18N2O Molecular Weight (g/mol): 218.3 MDL Number: MFCD08271925 InChI Key: KKYYCUZQPBFXKD-UHFFFAOYSA-N Synonym: 3-4-methylpiperazin-1-yl methyl benzaldehyde,benzaldehyde,3-4-methyl-1-piperazinyl methyl,3-4-methylpiperazinyl methyl benzaldehyde,3-4-methyl-1-piperazinyl methyl benzaldehyde PubChem CID: 18525774 IUPAC Name: 3-[(4-methylpiperazin-1-yl)methyl]benzaldehyde SMILES: CN1CCN(CC1)CC2=CC=CC(=C2)C=O