accessibility menu, dialog, popup

UserName

CAS RN 857284-25-4

NSHOFFF

CAS RN 857284-25-4

IUPAC Name: {2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}methanol
Synonyms: {2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}methanol
Molecular Weight (g/mol): 259.25
Molecular Formula: C11H8F3NOS
InChi Key: LXSRKDWWLYYTMN-UHFFFAOYSA-N
SMILES: OCC1=CSC(=N1)C1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results