accessibility menu, dialog, popup

UserName

CAS RN 85618-21-9

IUPAC Name: 2-(hydroxymethyl)-6-(octylsulfanyl)oxane-3,4,5-triol
Synonyms: 2-(hydroxymethyl)-6-(octylsulfanyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 308.43
Molecular Formula: C14H28O5S
InChi Key: CGVLVOOFCGWBCS-UHFFFAOYNA-N
SMILES: CCCCCCCCSC1OC(CO)C(O)C(O)C1O
Molecular Weight (g/mol) 
  • (2)
  • (6)
  • (3)
Quantity 
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (6)
  • (3)
Physical Form 
  • (6)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (6)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)