accessibility menu, dialog, popup

UserName

CAS RN 85553-44-2

BrS

CAS RN 85553-44-2

IUPAC Name: 2-[3-(bromomethyl)phenyl]thiophene
Synonyms: 2-[3-(bromomethyl)phenyl]thiophene
Molecular Weight (g/mol): 253.16
Molecular Formula: C11H9BrS
InChi Key: JQQDEBDYUAKNGD-UHFFFAOYSA-N
SMILES: BrCC1=CC=CC(=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results