accessibility menu, dialog, popup

UserName

CAS RN 854778-31-7

BFOCH3HOOH

CAS RN 854778-31-7

IUPAC Name: (4-fluoro-3-methoxyphenyl)boronic acid
Synonyms: 4-fluoro-3-methoxyphenylboronic acid
Molecular Weight (g/mol): 169.95
Molecular Formula: C7H8BFO3
InChi Key: LUJMSRVFSBMEOY-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results