accessibility menu, dialog, popup

UserName

CAS RN 854237-06-2

OHOBr

CAS RN 854237-06-2

IUPAC Name: 3'-bromo-[1,1'-biphenyl]-3-carboxylic acid
Synonyms: 3'-bromo-[1,1'-biphenyl]-3-carboxylic acid
Molecular Weight (g/mol): 277.12
Molecular Formula: C13H9BrO2
InChi Key: MHIAOGTWZISBLM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results