accessibility menu, dialog, popup

UserName

CH3SiH3CFOH

CAS RN 853955-70-1

IUPAC Name: {2-[(4-fluorophenyl)dimethylsilyl]phenyl}methanol
Synonyms: {2-[(4-fluorophenyl)dimethylsilyl]phenyl}methanol
Molecular Weight (g/mol): 260.38
Molecular Formula: C15H17FOSi
InChi Key: RKOHTBFNQRVSEP-UHFFFAOYSA-N
SMILES: C[Si](C)(C1=CC=C(F)C=C1)C1=CC=CC=C1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results