accessibility menu, dialog, popup

UserName

CAS RN 853771-91-2

H3COOHOFFF

CAS RN 853771-91-2

IUPAC Name: [4-methoxy-3-(trifluoromethoxy)phenyl]methanol
Synonyms: [4-methoxy-3-(trifluoromethoxy)phenyl]methanol
Molecular Weight (g/mol): 222.16
Molecular Formula: C9H9F3O3
InChi Key: XSWBFWDUPUANRL-UHFFFAOYSA-N
SMILES: COC1=C(OC(F)(F)F)C=C(CO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results