accessibility menu, dialog, popup

UserName

CAS RN 85326-10-9

IUPAC Name: N-[9-(5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl)-6-oxo-6,9-dihydro-3H-purin-2-yl]-2-methylpropanamide
Synonyms: N-[9-(5-{[(tert-butyldimethylsilyl)oxy]methyl}-4-hydroxyoxolan-2-yl)-6-oxo-3H-purin-2-yl]-2-methylpropanamide
Molecular Weight (g/mol): 451.6
Molecular Formula: C20H33N5O5Si
InChi Key: WZQWRHWLQPEBFF-UHFFFAOYNA-N
SMILES: CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C1CC(O)C(CO[Si](C)(C)C(C)(C)C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results