accessibility menu, dialog, popup

UserName

CAS RN 852180-53-1

BrSCl

CAS RN 852180-53-1

IUPAC Name: 5-bromo-3-(chloromethyl)-1-benzothiophene
Synonyms: 5-bromo-3-(chloromethyl)-1-benzothiophene
Molecular Weight (g/mol): 261.56
Molecular Formula: C9H6BrClS
InChi Key: ZIOALKCKGFEWMX-UHFFFAOYSA-N
SMILES: ClCC1=CSC2=CC=C(Br)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results