accessibility menu, dialog, popup

UserName

CAS RN 852180-39-3

NOOOHO

CAS RN 852180-39-3

IUPAC Name: 5-(pyridin-3-yloxy)furan-2-carboxylic acid
Synonyms: 5-(pyridin-3-yloxy)furan-2-carboxylic acid
Molecular Weight (g/mol): 205.17
Molecular Formula: C10H7NO4
InChi Key: ZTYMSWYWSCYFAF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OC2=CN=CC=C2)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results