accessibility menu, dialog, popup

UserName

CAS RN 851262-98-1

CH3OH3CFFF

CAS RN 851262-98-1

IUPAC Name: 1-[4-methyl-3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[4-methyl-3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 202.18
Molecular Formula: C10H9F3O
InChi Key: KXCYWGJWOOJKJP-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=C(C)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results