accessibility menu, dialog, popup

UserName

CAS RN 850589-56-9

BFOHOOH

CAS RN 850589-56-9

IUPAC Name: [3-(benzyloxy)-5-fluorophenyl]boronic acid
Synonyms: 3-(benzyloxy)-5-fluorophenylboronic acid
Molecular Weight (g/mol): 246.04
Molecular Formula: C13H12BFO3
InChi Key: QQOFKMPPQQMUDH-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(OCC2=CC=CC=C2)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results