accessibility menu, dialog, popup

UserName

BOHNOH3CHOOH

CAS RN 850568-17-1

IUPAC Name: [4-(methoxycarbamoyl)phenyl]boronic acid
Synonyms: 4-(methoxycarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 194.98
Molecular Formula: C8H10BNO4
InChi Key: MZZWCLBDQROHHS-UHFFFAOYSA-N
SMILES: CONC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results